72858
2-(2-Methyl-5-nitro-1H-imidazol-1-yl)acetic acid
analytical standard
Synonym(s):
1-Carboxymethylmetronidazole, 2-Methyl-5-nitroimidazole-1-acetic acid, Metronidazole-1-acetic acid
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥97.5% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
impurities
≤0.5% water
application(s)
forensics and toxicology
pharmaceutical (small molecule)
format
neat
SMILES string
O=C(O)CN1C(C)=NC=C1[N+]([O-])=O
InChI
1S/C6H7N3O4/c1-4-7-2-5(9(12)13)8(4)3-6(10)11/h2H,3H2,1H3,(H,10,11)
InChI key
RVEGZXNRNWUYKI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service