Skip to Content
Merck

Skip To

72015

Sodium tetrakis[3,5-bis(1,1,1,3,3,3-hexafluoro-2-methoxy-2-propyl)phenyl]borate trihydrate

Selectophore

Synonym(s):

HFPB, Tetrakis[3,5-bis(1,1,1,3,3,3-hexafluoro-2-methoxy-2-propyl)phenyl]boron sodium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 mg
Check Cart for Availability
€ 254,00

About This Item

Empirical Formula (Hill Notation):
C56H36BF48NaO8 · 3H2O
CAS Number:
Molecular Weight:
1836.65
NACRES:
NB.61
PubChem Substance ID:
UNSPSC Code:
26111700
MDL number:

€ 254,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

for ion-selective electrodes

Quality Segment

product line

Selectophore

form

powder

SMILES string

O.O.O.[Na+].COC(c1cc(cc(c1)C(OC)(C(F)(F)F)C(F)(F)F)[B-](c2cc(cc(c2)C(OC)(C(F)(F)F)C(F)(F)F)C(OC)(C(F)(F)F)C(F)(F)F)(c3cc(cc(c3)C(OC)(C(F)(F)F)C(F)(F)F)C(OC)(C(F)(F)F)C(F)(F)F)c4cc(cc(c4)C(OC)(C(F)(F)F)C(F)(F)F)C(OC)(C(F)(F)F)C(F)(F)F)(C(F)(F)F)C(F)(F)F

InChI

1S/C56H36BF48O8.Na.3H2O/c1-106-33(41(58,59)60,42(61,62)63)21-9-22(34(107-2,43(64,65)66)44(67,68)69)14-29(13-21)57(30-15-23(35(108-3,45(70,71)72)46(73,74)75)10-24(16-30)36(109-4,47(76,77)78)48(79,80)81,31-17-25(37(110-5,49(82,83)84)50(85,86)87)11-26(18-31)38(111-6,51(88,89)90)52(91,92)93)32-19-27(39(112-7,53(94,95)96)54(97,98)99)12-28(20-32)40(113-8,55(100,101)102)56(103,104)105;;;;/h9-20H,1-8H3;;3*1H2/q-1;+1;;;

InChI key

HQYSIIBSDLWUEX-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Lipophilic ion exchanger for ion-selective membrane electrodes; Lipophilic salt as additive in membrane electrodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
605917201424901
description

for ion-selective electrodes

description

for ion-selective electrodes

description

for ion-selective electrodes

description

for ion-selective electrodes

product line

Selectophore

product line

Selectophore

product line

Selectophore

product line

Selectophore

form

powder

form

solid

form

solid

form

powder

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters