Skip to Content
Merck
All Photos(1)

Key Documents

71878

Sigma-Aldrich

H-D-Lys(Alloc)-OH

puriss., ≥99.0% (HPLC)

Synonym(s):

Nδ-Alloc-D-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18N2O4
CAS Number:
Molecular Weight:
230.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

puriss.

Assay

≥99.0% (HPLC)

form

lumps

optical activity

[α]20/D −18.5±1°, c = 1% in 1 M HCl

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

N[C@H](CCCCNC(=O)OCC=C)C(O)=O

InChI

1S/C10H18N2O4/c1-2-7-16-10(15)12-6-4-3-5-8(11)9(13)14/h2,8H,1,3-7,11H2,(H,12,15)(H,13,14)/t8-/m1/s1

InChI key

JMZPOGGPUVRZMI-MRVPVSSYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service