Skip to Content
Merck
All Photos(1)

Key Documents

71409

Sigma-Aldrich

Sodium citrate tribasic dihydrate

tested according to Ph. Eur.

Synonym(s):

Natrii citras, Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COONa)(CH2COONa)2 · 2H2O
CAS Number:
Molecular Weight:
294.10
Beilstein:
6104939
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

Assay

99-101% anhydrous basis

form

powder

mp

>300 °C (lit.)

solubility

water: very soluble 29.4 g/L at 20 °C

SMILES string

O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Na.2H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;2*1H2/q;3*+1;;/p-3

InChI key

NLJMYIDDQXHKNR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Zhao et al.
Nanotechnology, 25(40), 405101-405101 (2014-09-12)
The treatment of liver injuries or hepatocellular carcinoma (HCC) has been hindered by the lack of efficient drug delivery. Even with the help of nanoparticles or other synthetic delivering agents, a large portion of the dose is still sequestered in
Ningning He et al.
Molecular cancer therapeutics, 13(10), 2463-2473 (2014-08-15)
The recent proliferation of data on large collections of well-characterized cancer cell lines linked to therapeutic drug responses has made it possible to identify lineage- and mutation-specific transcriptional markers that can help optimize implementation of anticancer agents. Here, we leverage
Héctor Rodríguez-Rodríguez et al.
Small (Weinheim an der Bergstrasse, Germany), 11(13), 1555-1561 (2014-12-03)
3D remote control of multifunctional fluorescent up-converting nanoparticles (UCNPs) using optical forces is being required for a great variety of applications including single-particle spectroscopy, single-particle intracellular sensing, controlled and selective light-activated drug delivery and light control at the nanoscale. Most
Yan-Ming Liu et al.
Colloids and surfaces. B, Biointerfaces, 122, 287-293 (2014-07-30)
A label-free and ultrasensitive electrochemiluminescence (ECL) aptasensor was constructed for the detection of thrombin. Molybdenum sulfide-graphene nanocomposites with good conductivity and large surface area were immobilized on glassy carbon electrode (GCE), and then Nafion was fixed to chemosorb the Ru(bpy)3(2+)
Wenchao Ding et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 16(2), 447-454 (2014-11-14)
The Turkevich method, involving the reduction of HAuCl4 with citrate in boiling water, allows the facile production of monodisperse, quasispherical gold nanoparticles (AuNPs). Although, it is well-known that the size of the AuNPs obtained with the same recipe vary slightly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service