Skip to Content
Merck
All Photos(1)

Key Documents

70961

Supelco

TOC standard

according to ISO/CEN EN1484, 100 mg/L±5 mg/L

Synonym(s):

Total Organic Carbon Reference Material

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€116.00
500 ML
€221.00

€116.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€116.00
500 ML
€221.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€116.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Agency

according to ISO/CEN EN1484

quality

stabilized

shelf life

limited shelf life, expiry date on the label

concentration

100 mg/L±5 mg/L

technique(s)

TOC: suitable

suitability

in accordance for application

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Total organic carbon (TOC) is a measure of the total amount of carbon in an organic compound. It is a critical parameter to evaluate the water quality for possible organic contamination.

Application

This analytical standard is intended for the calibration and validation of analytical instruments used to measure the total organic carbon (TOC) of drinking water, groundwater, surface water, seawater, and wastewater in accordance with ISO/CEN EN1484.

TOC standard may be used in the analysis of organic matter, which determines the water quality using ultraviolet-visible spectrophotometric technique.[1]

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helene Baribeau, AWWA Research Foundation
Formation and Decay of Disinfection By-products in the Distribution System (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service