Skip to Content
Merck
All Photos(1)

Key Documents

70905

Supelco

Palm oil

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
NA.21
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

quality

refined

composition

linoleic acid, 6-13% (fatty acid)
myristic acid, 0.5-6% (fatty acid)
oleic acid, 35-50% (fatty acid)
palmitic acid, 35-48% (fatty acid)
stearic acid, 3-7% (fatty acid)

mp

30-40 °C (lit.)

acid value

≤0.4

iodine value

50‑57

saponification value

190‑210

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Mountapmbeme-Kouotou et al.
Environmental monitoring and assessment, 185(7), 5789-5800 (2012-12-12)
The gliding electric discharge in humid air is a source of activated species forming (e.g. (•)OH, (•)NO and their derivatives H2O2, ONO2H and NO3H) which are present in a non-thermal plasma at atmospheric pressure. These species are able to degrade
Joaquín C Surra et al.
The British journal of nutrition, 109(2), 202-209 (2013-01-11)
Epidemiological studies have demonstrated the benefits of nut consumption on cardiovascular risk factors and CHD, attributed to their fatty acid profile, rich in unsaturated fatty acids, and also to other nutrients. The effect of nuts on atherosclerotic lesions was studied
Béatrice Gleize et al.
The British journal of nutrition, 110(1), 1-10 (2012-12-12)
The xanthophylls lutein and zeaxanthin probably play a role in visual function and may participate in the prevention of age-related eye diseases. Although a minimum amount of TAG is required for an optimal bioavailability of these carotenoids, the effect of
Xia Zhang et al.
Molecules (Basel, Switzerland), 18(1), 1036-1052 (2013-01-17)
Thermal behavior of palm stearin (PS) and palm olein (PO) was explored by monitoring peak temperature transitions by differential scanning calorimetry (DSC). The fatty acid composition (FAC), isothermal crystallization kinetics studied by pulsed Nuclear Magnetic Resonance (pNMR) and isothermal microstructure
Forests: Oil-palm concerns in Brazilian Amazon.
Alexander C Lees et al.
Nature, 497(7448), 188-188 (2013-05-10)

Questions

  1. Dear all, could you please state for the research proposes, if the product Palm oil no. 70905, refined, is composed only out of free fatty acids or the fatty acids are in the triglyceride form? Thank you for your reply in advance.

    1 answer
    1. The product is comprised primarily of free fatty acids, with the majority identified as Palmitic, Oleic, Linoleic, Stearic, and Myristic acids. There may be residual triglycerides in trace amounts, however, no additional testing is performed to further confirm or identify these components. The method of manufacturing is considered proprietary. The analytical data does suggest that the product has been refined to some degree.
      Please see the link below to review a sample Certificate of Analysis:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/70905-BULKBCCK5908.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service