Skip to Content
Merck
All Photos(1)

Key Documents

70193

Millipore

Staphylococcus Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

CHAPMAN-Agar, Staphylococcus Medium No. 110

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€93.60

€93.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€93.60

About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

€93.60


Check Cart for Availability

Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
dipotassium hydrogen phosphate, 5.0 g/L
gelatin, 30.0 g/L
lactose, 2.0 g/L
mannitol, 10.0 g/L
sodium chloride, 75.0 g/L
tryptone, 10.0 g/L
yeast extract, 2.5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.1±0.2 (25 °C)

application(s)

bioburden testing
clinical testing
environmental
food and beverages
veterinary

microbiology

suitability

slightly selective and differential for Staphylococcus spp.

Application

Selective medium for isolation and differentiation of pathogenic staphylococci on the basis of salt tolerance, pigmentation, D-mannitol utilization and gelatin liquefaction. Smuckler and Appleman recommend addition of sodium azide in order to make this medium selective for the determination of coagulase-positive staphylococci.

Preparation Note

Dissolve 149.5 g in 1 litre distilled water and sterilize by autoclaving at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple method for making multiple tests of a micro-organism.
G H CHAPMAN
Journal of bacteriology, 63(1), 147-149 (1952-01-01)
R P COLLINS et al.
Applied microbiology, 12, 335-336 (1964-07-01)
By use of various trapping systems, as well as lead acetate papers, Streptomyces odorifer was shown to produce hydrogen sulfide. Other sulfur-containing compounds may be produced by S. odorifer, but the amounts obtained were too small for detailed analysis. It

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service