Skip to Content
Merck
All Photos(1)

Documents

68982

Supelco

Methyl 2-octynoate

analytical standard

Synonym(s):

Methyl heptine carbonate, NSC 72098

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4C≡CCO2CH3
CAS Number:
Molecular Weight:
154.21
Beilstein:
1756887
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.446 (lit.)
n20/D 1.446-1.448

bp

217-220 °C (lit.)

density

0.92 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCC#CC(=O)OC

InChI

1S/C9H14O2/c1-3-4-5-6-7-8-9(10)11-2/h3-6H2,1-2H3

InChI key

FRLZQXRXIKQFNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Skin Sens. 1A

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service