Skip to Content
Merck
All Photos(2)

Documents

68535

Supelco

Halofenozide

PESTANAL®, analytical standard

Synonym(s):

N-tert-Butyl-N′-(4-chlorobenzoyl)benzohydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19ClN2O2
CAS Number:
Molecular Weight:
330.81
Beilstein:
8421868
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

composition

carbon content, 65.35%
hydrogen content, 5.79%
nitrogen content, 8.47%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)(C)N(NC(=O)c1ccc(Cl)cc1)C(=O)c2ccccc2

InChI

1S/C18H19ClN2O2/c1-18(2,3)21(17(23)14-7-5-4-6-8-14)20-16(22)13-9-11-15(19)12-10-13/h4-12H,1-3H3,(H,20,22)

InChI key

CNKHSLKYRMDDNQ-UHFFFAOYSA-N

General description

Halofenozide is a systemic type of insect growth regulator belonging to the class of non-steroidal ecdysone agonists. It shows a broad spectrum of activity in controlling beetle grub and lepidopteran larval pests.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of ecdysone agonist halofenozide against Culex pipiens.
Boudjelida H, et al.
Pesticide Biochemistry and Physiology, 83(2-3), 115-123 (2005)
Rebekah A Reynolds et al.
Scientific reports, 10(1), 21084-21084 (2020-12-05)
Mosquito physiology and immunity are integral determinants of malaria vector competence. This includes the principal role of hormonal signaling in Anopheles gambiae initiated shortly after blood-feeding, which stimulates immune induction and promotes vitellogenesis through the function of 20-hydroxyecdysone (20E). Previous
Insect Control: Biological and Synthetic Agents (2010)
Ji-Feng Shi et al.
Insect biochemistry and molecular biology, 68, 1-12 (2015-11-26)
Uridine diphosphate-N-acetylglucosamine-pyrophosphorylase (UAP) is involved in the biosynthesis of chitin, an essential component of the epidermal cuticle and midgut peritrophic matrix (PM) in insects. In the present paper, two putative LdUAP genes were cloned in Leptinotarsa decemlineata. In vivo bioassay revealed
Jinhua Gan et al.
Ecotoxicology and environmental safety, 210, 111853-111853 (2021-01-11)
A multi-residue analysis of six diacylhydrazine insecticides in water, sediment, and aquatic products was established by liquid chromatography triple quadrupole tandem mass spectrometry (LC-MS/MS). The water sample was extracted with acetonitrile by low-temperature enrichment liquid-liquid extraction technology. The sediment and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service