Skip to Content
Merck
All Photos(2)

Key Documents

68459

Supelco

Ethosuximide

analytical standard

Synonym(s):

2-Ethyl-2-methylsuccinimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NO2
CAS Number:
Molecular Weight:
141.17
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

O=C1NC(CC1(C)CC)=O

InChI

1S/C7H11NO2/c1-3-7(2)4-5(9)8-6(7)10/h3-4H2,1-2H3,(H,8,9,10)

InChI key

HAPOVYFOVVWLRS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E B Posner et al.
The Cochrane database of systematic reviews, (3)(3), CD003032-CD003032 (2003-08-15)
Absence seizures are brief epileptic seizures which present in childhood and adolescence. They are characterised by sudden loss of awareness and an electroencephalogram (EEG) typically shows generalised spike wave discharges at three cycles per second. Ethosuximide, valproate and lamotrigine are
E B Posner et al.
The Cochrane database of systematic reviews, (4)(4), CD003032-CD003032 (2005-10-20)
Absence seizures are brief epileptic seizures which present in childhood and adolescence. They are characterised by sudden loss of awareness and an electroencephalogram (EEG) typically shows generalised spike wave discharges at three cycles per second. Ethosuximide, valproate and lamotrigine are
G V Massey et al.
Pediatric neurology, 11(1), 59-61 (1994-07-01)
We describe a case of aplastic anemia in an 8-year-old girl which was diagnosed 8 months after initiation of ethosuximide as treatment for absence seizures. Blood counts had been previously monitored and were normal. The patient successfully underwent allogeneic bone
[Ethosuximide].
M Ichiba et al.
Nihon rinsho. Japanese journal of clinical medicine, 53 Su Pt 1, 915-917 (1995-02-01)
J S Millership et al.
European journal of drug metabolism and pharmacokinetics, 18(4), 349-353 (1993-10-01)
The metabolism of the antiepileptic drug ethosuximide (3-ethyl-3-methylpyrollidine-2,5-dione) (I) in animals and humans is reviewed. Chiral aspects of the metabolism of ethosuximide are discussed. Clarification of the precise nature of the hydroxymetabolites of ethosuximide is presented.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service