Skip to Content
Merck
All Photos(1)

Key Documents

68217

Supelco

2-Oxobutyric acid

analytical standard

Synonym(s):

α-Ketobutyric acid, 2-Oxobutanoic acid, Propionylformic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COCOOH
CAS Number:
Molecular Weight:
102.09
Beilstein:
1700514
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

96.5-103.5% (T)

shelf life

limited shelf life, expiry date on the label

mp

30-33 °C

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCC(C(O)=O)=O

InChI

1S/C4H6O3/c1-2-3(5)4(6)7/h2H2,1H3,(H,6,7)

InChI key

TYEYBOSBBBHJIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Oxobutyric acid, an α-keto acid, is an important intermediate in the isoleucine fermentation metabolic pathway. It is regarded as a key raw material for chiral 2-aminobutyric and 2-hydroxybutyric acids and paulomycin production.

Application

2-Oxobutyric acid (α-ketobutyric acid) may be used as an analytical reference standard for the determination of the analyte in K562 cells by high-performance liquid chromatography with fluorescence detection and in drinking water at low microgram per liter concentrations by ion chromatography technique.

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of short-chain aliphathic, oxo-and hydroxy-acids in drinking water at low microgram per liter concentrations
Peldszus S, et al.
Journal of Chromatography A, 723(1), 27-34 (1996)
Analysis of intracellular $\alpha$-keto acids by HPLC with fluorescence detection
Fujiwara T, et al.
Analytical Methods : Advancing Methods and Applications, 12(20), 2555-2559 (2020)
Microbial production of 2-oxobutyric acid from crotonic acid
Furuyoshi S, et al.
Agricultural and Biological Chemistry, 55(1), 123-128 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service