Skip to Content
Merck
All Photos(1)

Documents

68113

Supelco

2-{1-[(2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-4-[2-(3-methoxyphenyl)-2-oxoethylthio]piperidin-3-ylidene}acetic acid hydrochloride

analytical standard

Synonym(s):

Prasugrel metabolite M3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H28FNO5S · HCl
CAS Number:
Molecular Weight:
534.04
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Assay

≥97.0% (sum of diastereomers, HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

storage temp.

−20°C

SMILES string

O=C(C1CC1)C(N2CCC(SCC(C3=CC(OC)=CC=C3)=O)/C(C2)=C\C(O)=O)C4=CC=CC=C4F.Cl

InChI

1S/C27H28FNO5S.ClH/c1-34-20-6-4-5-18(13-20)23(30)16-35-24-11-12-29(15-19(24)14-25(31)32)26(27(33)17-9-10-17)21-7-2-3-8-22(21)28;/h2-8,13-14,17,24,26H,9-12,15-16H2,1H3,(H,31,32);1H/b19-14-;

InChI key

IXPJVHUXCREIQM-YEBWQKSTSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service