Skip to Content
Merck
All Photos(1)

Key Documents

67861

Supelco

DL-3-(4-Hydroxyphenyl)lactic acid

analytical standard

Synonym(s):

DL-p-Hydroxyphenyllactic acid, 3-(4-Hydroxyphenyl)-2-hydroxypropanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€74.50

€74.50


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€74.50

About This Item

Empirical Formula (Hill Notation):
C9H10O4
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.24

€74.50


Estimated to ship on21 May 2025


Request a Bulk Order

grade

analytical standard

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤10% water (calc. from elemental analysis)

mp

137-143 °C

SMILES string

OC(Cc1ccc(O)cc1)C(O)=O

InChI

1S/C9H10O4/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4,8,10-11H,5H2,(H,12,13)

InChI key

JVGVDSSUAVXRDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service