Skip to Content
Merck
All Photos(2)

Key Documents

67805

Supelco

Idebenone

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€66.60

€66.60


Estimated to ship on04 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€66.60

About This Item

Empirical Formula (Hill Notation):
C19H30O5
CAS Number:
Molecular Weight:
338.44
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€66.60


Estimated to ship on04 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

O=C(C(OC)=C1OC)C(CCCCCCCCCCO)=C(C)C1=O

InChI

1S/C19H30O5/c1-14-15(12-10-8-6-4-5-7-9-11-13-20)17(22)19(24-3)18(23-2)16(14)21/h20H,4-13H2,1-3H3

InChI key

JGPMMRGNQUBGND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

A short-chain Coenzyme Q analog that enhances superoxide formation, presumably by mediating electron transfer from N2 to oxygen.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael H Parkinson et al.
Journal of neurochemistry, 126 Suppl 1, 125-141 (2013-07-24)
Friedreich's ataxia is a debilitating progressive neurodegenerative disease associated with cardiomyopathy and other features. The underlying cause is a deficiency of the mitochondrial protein frataxin which causes mitochondrial iron deposition, increased oxidative stress and impaired adenosine triphosphate production. Over the
[The use of idebenone (noben) in neurological practice].
I V Damulin
Zhurnal nevrologii i psikhiatrii imeni S.S. Korsakova, 106(11), 66-71 (2006-12-21)
Idebenone - monograph.
Alternative medicine review : a journal of clinical therapeutic, 6(1), 83-86 (2001-02-24)
Claudia Carbone et al.
Expert opinion on drug delivery, 9(11), 1377-1392 (2012-10-23)
Idebenone (IDE) is an antioxidant compound, structurally related to coenzyme Q10. Its therapeutic potential is growing in different application areas, as demonstrated by the number of experimental works and patents produced in very recent years. Cyclodextrin inclusion complexes, liposomes, microemulsions
Jörg B Schulz et al.
Journal of neurology, 256 Suppl 1, 42-45 (2009-04-11)
Several reports in the literature describe the effects of low-dose (5 mg/kg/day) idebenone in significantly reducing cardiac hypertrophy in patients with Friedreich ataxia. However, the effects of idebenone on neurological function have not been reliably determined in these studies; when

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service