Skip to Content
Merck
All Photos(1)

Documents

67334

Supelco

Diphenyl ether

Selectophore, ≥99.9%

Synonym(s):

Diphenyl oxide, Phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2O
CAS Number:
Molecular Weight:
170.21
Beilstein:
1364620
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

vapor density

>5.86 (25 °C, vs air)

vapor pressure

<1 mmHg ( 20 °C)

product line

Selectophore

Assay

≥99.9% (GC)
≥99.9%

form

solid

autoignition temp.

1144 °F

expl. lim.

1.5 %

refractive index

n20/D 1.579 (lit.)

bp

259 °C (lit.)

mp

25-27 °C (lit.)
27-29 °C

density

1.073 g/mL at 25 °C (lit.)

SMILES string

O(c1ccccc1)c2ccccc2

InChI

1S/C12H10O/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10H

InChI key

USIUVYZYUHIAEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diphenyl ether (diphenyl oxide) belongs to the family of aromatic homomonocyclic compounds. It is colorless, exists as crystalline solid or liquid (above 82°F) and has a geranium-like odor. Diphenyl ether is present in alcoholic beverages. It is used as a flavouring ingredient and is found in vanilla, green tea, muscat grapes, potato chips, grilled beef, buckwheat, and lemon balm. It is a starting material in the production of phenoxathiin via the Ferrario reaction.. In similar reactions, diphenyl ether is a significant side product in the high-pressure hydrolysis of chlorobenzene during production of phenol. Also several polybrominated diphenyl ethers (PBDEs) are useful flame retardants.
Visit our Sensor Applications portal to learn more.

Application

Used as plasticizing solvent mediator for PVC-based membranes.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chandra, S., Lang, H.
Sensors and Actuators B, Chemical, 114, 849-854 (2006)
Kostantinos Benekos et al.
Journal of biotechnology, 150(1), 195-201 (2010-07-20)
Plant glutathione transferases (GSTs) superfamily consists of multifunctional enzymes and forms a major part of the plants herbicide detoxification enzyme network. The tau class GST isoenzyme GmGSTU4 from soybean, exhibits catalytic activity towards the diphenyl ether herbicide fluorodifen and is
Ge-Fei Hao et al.
Journal of computer-aided molecular design, 25(3), 213-222 (2011-01-25)
Protoporphyrinogen oxidase (PPO, EC 1.3.3.4), which has been identified as a significant target for a great family of herbicides with diverse chemical structures, is the last common enzyme responsible for the seventh step in the biosynthetic pathway to heme and
Ariana Beste et al.
The Journal of organic chemistry, 74(7), 2837-2841 (2009-03-06)
Lignin is an abundant natural resource that is a potential source of valuable chemicals. Improved understanding of the pyrolysis of lignin occurs through the study of model compounds for which phenethyl phenyl ether (PhCH(2)CH(2)OPh, PPE) is the simplest example representing
Tony Ly et al.
Analytical chemistry, 80(13), 5059-5064 (2008-05-24)
Selective noncovalent adduct protein probing (SNAPP) mass spectrometry was recently developed to study solution-phase conformations of proteins by exploiting the specific affinity between 18-crown-6 ether (18C6) and lysine side chains. To obtain more detailed information about protein tertiary structure, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service