Skip to Content
Merck
All Photos(2)

Key Documents

66681

Supelco

4,4′-Methylene-bis(2-chloroaniline)

analytical standard

Synonym(s):

2,2′-Dichloro-4,4′-methylenedianiline, 4,4′-Diamino-3,3′-dichlorodiphenylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2[C6H3(Cl)NH2]2
CAS Number:
Molecular Weight:
267.15
Beilstein:
1882318
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

202-214 °C/0.3 mmHg (lit.)

mp

102-107 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Nc1ccc(Cc2ccc(N)c(Cl)c2)cc1Cl

InChI

1S/C13H12Cl2N2/c14-10-6-8(1-3-12(10)16)5-9-2-4-13(17)11(15)7-9/h1-4,6-7H,5,16-17H2

InChI key

IBOFVQJTBBUKMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-Methylene-bis(2-chloroaniline) is an azo dye metabolite that is deemed toxic to the environment.

Application

4,4′-Methylene-bis(2-chloroaniline) may be used as an analytical reference standard for the determination of 4,4′-methylene-bis(2-chloroaniline) in:
  • Urine samples by high performance liquid chromatography (HPLC).
  • Textile samples by HPLC coupled to tandem mass spectrometry (MS/MS) equipped with multiple reaction monitoring (MRM) mode of detection.
  • Plastic multilayer food packaging materials by LC-Orbitrap-full scan-high resolution mass spectrometry (HRMS) equipped with electrospray ionization (ESI) source.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Standard for determining aromatic amines released from azo-dyes in commodities

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Rohm et al.
GIT Labor-Fachzeit., 1080-1080 (1997)
Degradability of selected azo dye metabolites in activated sludge systems.
Ekici P, et al.
Chemosphere, 44(4), 721-728 (2001)
Evaluation of worker exposure to TDI, MOCA, and methylene chloride.
Richard Fairfax et al.
Journal of occupational and environmental hygiene, 3(6), D50-D53 (2006-04-20)
P Vineis et al.
Cancer causes & control : CCC, 8(3), 346-355 (1997-05-01)
Epidemiological evidence on the relation between aromatic amines and cancer risk is reviewed. In particular, cancer risk in humans resulting from exposure to aromatic amines from occupational sources and tobacco smoking is assessed with reference to ecologic, cohort, and case-control
Liquid chromatography-full scan-high resolution mass spectrometry-based method towards the comprehensive analysis of migration of primary aromatic amines from food packaging.
Mattarozzi M, et al.
Journal of Chromatography A, 1320(1), 96-102 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service