Skip to Content
Merck
All Photos(1)

Key Documents

64120

Sigma-Aldrich

Methacryloyl chloride

purum, dist., ≥97.0% (GC), contains ~0.02% 2,6-di-tert-butyl-4-methylphenol as stabilizer

Synonym(s):

2-Methylprop-2-enoyl chloride, Methacrylic acid chloride, Methacrylyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COCl
CAS Number:
Molecular Weight:
104.53
Beilstein:
878175
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

purum

Quality Level

Assay

≥97.0% (GC)

quality

dist.

contains

~0.02% 2,6-di-tert-butyl-4-methylphenol as stabilizer

impurities

≤1000 mg/kg total sulfur (as SO4)

refractive index

n20/D 1.442 (lit.)
n20/D 1.444

bp

95-96 °C (lit.)

density

1.08 g/mL at 20 °C
1.07 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(Cl)=O

InChI

1S/C4H5ClO/c1-3(2)4(5)6/h1H2,2H3

InChI key

VHRYZQNGTZXDNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methacryloyl chloride is the acid chloride of methacrylic acid that is used in the field of polymers as a monomer or a reactive intermediate in the synthesis of various polymers. It is used to synthesize various polymers, such as poly(methacryloyl chloride), poly(methacrylates), and poly(methacrylamides) for wide applications including the production of dental materials, adhesives, contact lenses, hydrogels, coatings, and in the development of various biomedical devices.

Application

Methacryloyl chloride can be used as:
  • A monomer in the synthesis of amphiphilic fluorescent copolymers via reversible addition-fragmentation chain transfer (RAFT) polymerization. The resulting copolymers have smart pH sensitivity and fluorescence properties, which make them suitable for use in cell imaging.
  • A monomer in the emulsion polymerization process to prepare a fluorescence probe, which is used for the detection of metronidazole.
  • A surface modifier for microcrystalline cellulose (MCC) to improve its compatibility with low-density polyethylene (LDPE) in the preparation of LDPE-based composites.
  • A key component in the synthesis of carboxymethyl cellulose acrylates, which find applications in the development of pH-sensitive hydrogels.
  • A monomer in the synthesis of methacrylate-based hydrogel materials, which are widely used in the production of soft contact lenses.
Methacryloyl chloride can also be used:
  • To prepare methacryloyl-functional benzoxazine monomer.
  • As a precursor to prepare N-methacryloyl-(L)-histidinemethylester(MAH), a metal chelating monomer.

replaced by

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.0 °F - closed cup

Flash Point(C)

12.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Runhai Chen et al.
Macromolecular bioscience, 20(2), e1900331-e1900331 (2019-12-20)
Polymer nanoparticulate drug delivery systems that respond to reactive oxygen species (ROS) and glutathione (GSH) simultaneously at biologically relevant levels hold great promise to improve the therapeutic efficacy to cancer cells with reduced side effects of chemo drugs. Herein, a
Xiangzheng Hu et al.
Steroids, 70(8), 531-537 (2005-05-17)
Bile acids in the family of steroid compounds can be chemically modified for biochemical and other applications. Derivatives of cholic acid with multiple methacrylate groups can be prepared by the use of methacrylic acid, methacryloyl chloride and methacryloyl anhydride as
E Asmussen et al.
Scandinavian journal of dental research, 91(2), 153-155 (1983-04-01)
The bonding of restorative resins to dentin by means of an intermediary monomer containing a carboxylic acid chloride or an isocyanate group was investigated. Such intermediary monomers are supposed to react with the organic constituent of dentin. Bond strengths were
Polymerized phosphatidyl choline vesicles. Stabilized and controllable time-release carriers.
S L Regen et al.
Biochemical and biophysical research communications, 101(1), 131-136 (1981-07-16)
Jinghui Li et al.
Scientific reports, 7, 40082-40082 (2017-01-05)
Crosslinking is an effective way to fabricate high-selective CO

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service