Skip to Content
Merck
All Photos(1)

Documents

63961

Supelco

Triethylamine hydrochloride

suitable for HPLC, LiChropur, 99.0-101.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3N · HCl
CAS Number:
Molecular Weight:
137.65
Beilstein:
3906409
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.21

Assay

99.0-101.0% (AT)

form

solid

technique(s)

HPLC: suitable

impurities

≤1.0% water

mp

256-259 °C
261 °C (dec.) (lit.)

solubility

H2O: 1 g/10 mL, clear, colorless

λ

in H2O (3 g in 30 mL)

UV absorption

λ: 220 nm Amax: ≤0.05
λ: 230 nm Amax: ≤0.04
λ: 250 nm Amax: ≤0.03
λ: 260 nm Amax: ≤0.02
λ: 500 nm Amax: ≤0.02

SMILES string

Cl.CCN(CC)CC

InChI

1S/C6H15N.ClH/c1-4-7(5-2)6-3;/h4-6H2,1-3H3;1H

InChI key

ILWRPSCZWQJDMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service