Skip to Content
Merck
All Photos(1)

Key Documents

61031BC

Millipore

Assurance® GDS Wash Solution PCR Reagent

For use with Assurance® GDS

Synonym(s):

Wash Solution for PCR Sample Prep

Sign Into View Organizational & Contract Pricing

Select a Size

4 X 250 ML
€166.00

€166.00


Estimated to ship on24 May 2025


Request more information

Select a Size

Change View
4 X 250 ML
€166.00

About This Item

UNSPSC Code:
41106306
NACRES:
NA.85

€166.00


Estimated to ship on24 May 2025


Request more information

Product Name

GDS Wash Solution Kit, BioControl, For use with GDS

Quality Level

form

liquid

shelf life

6‑12 mo.

packaging

bottle of 4 × 250 mL each

manufacturer/tradename

BioControl Systems 61031-100

storage condition

(Tightly closed in a dry, cool, and well-ventilated place.)

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with BioControl GDS

storage temp.

2-8°C

suitability

Escherichia coli (O157:H7)
Cronobacter spp.
Listeria spp.
Salmonella spp.

General description

GDS Wash Solution Kit is recommended to use with Assurance® GDS assays. Assurance® GDS is an automated nucleic acid amplification system for the detection of pathogens. This larger size format is commonly purchased with automation kits for use with Assurance® GDS PickPen® PipetMax® (PPMX).

Application

GDS Wash Solution Kit is used during the sample preparation for the detection of pathogens in foods and environmental samples using Assurance® GDS kits.

Components

Four bottles, 250 mL each

Testing Method

PCR

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
BioControl is a registered trademark of BioControl Systems, Inc.
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc

related product

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service