Skip to Content
Merck
All Photos(1)

Key Documents

61030BC

Millipore

GDS Poly IMS - Top STEC

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€472.00

€472.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 EACH
€472.00

About This Item

UNSPSC Code:
41106302
NACRES:
HA.03

€472.00


Check Cart for Availability

Request more information

shelf life

6-12 mo.

Quality Level

technique(s)

PCR: suitable

application(s)

food and beverages

compatibility

BioControl GDS

storage temp.

2-8°C

suitability

enterovirulent E. coli (EEC) spp.

General description

Assurance® GDS Poly IMS - Top STEC contains a mixture of antibody-coated paramagnetic particles targeted against the Top 7 serogroups of Shiga Toxigenic E. coli (STEC).

Application

The reagent can be utilized to help confirm STEC positive samples by isolating organisms belonging to O-serogroups O103, O111, O121, O145, O26, O45 and O157 from the positive enrichment broth.

Components

One bottle, 2.4 mL

Testing Method

PCR

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Assurance® GDS offers integrated solution for fast and accurate food pathogen detection in microbiology testing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service