Skip to Content
Merck
All Photos(1)

Key Documents

59974

Supelco

2-Hydroxy-3-[4-(2-methoxyethyl)phenoxy]propionic acid

analytical standard

Synonym(s):

2-Hydroxy-3-[4-(2-methoxyethyl)phenoxy]propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16O5
CAS Number:
Molecular Weight:
240.25
Beilstein:
25973382
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(C(O)=O)COC1=CC=C(CCOC)C=C1

InChI

1S/C12H16O5/c1-16-7-6-9-2-4-10(5-3-9)17-8-11(13)12(14)15/h2-5,11,13H,6-8H2,1H3,(H,14,15)

InChI key

PQIWJWWULVHOAA-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service