Skip to Content
Merck
All Photos(1)

Key Documents

55822

Supelco

Ginkgolic acid C17:1

analytical standard

Synonym(s):

6-[(10Z)-Heptadecenyl]salicylic acid, Ginkgolic acid II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H38O3
CAS Number:
Molecular Weight:
374.56
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CCCCCC\C=C/CCCCCCCCCc1cccc(O)c1C(O)=O

InChI

1S/C24H38O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-18-21-19-17-20-22(25)23(21)24(26)27/h7-8,17,19-20,25H,2-6,9-16,18H2,1H3,(H,26,27)/b8-7-

InChI key

MBYNDKVOZOAOIS-FPLPWBNLSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ginkgolic acid C17:1 is a toxic phenolic compound obtained from the leaves and fruits of Ginkgo biloba.[1][2]

Application

Ginkgolic acid C17:1 may be used as a reference standard for the determination of ginkgolic acid C17:1 content in Ginkgo biloba leaves using high-performance liquid chromatography (HPLC).[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Determination of ginkgolic acids by high performance liquid chromatography.
QLY, et al.
Yao Xue Xue Bao = Acta Pharmaceutica Sinica, 37(7), 555-558 (2002)
Muhammad Nadeem Akhtar et al.
Bioorganic & medicinal chemistry letters, 21(13), 4097-4103 (2011-06-07)
Bioassay-guided extraction of the stem bark of Knema laurina showed the acetylcholinesterase (AChE) inhibitory activity of DCM and hexane fractions. Further repeated column chromatography of hexane and DCM fractions resulted in the isolation and purification of five alkenyl phenol and
Edible Medicinal And Non Medicinal Plants (2012)
Edible Medicinal And Non Medicinal Plants (2012)
K Ndjoko et al.
Journal of chromatography. B, Biomedical sciences and applications, 744(2), 249-255 (2000-09-19)
Ginkgolic acids (GAs) are toxic phenolic compounds present in the fruits and leaves of Ginkgo biloba L. (Ginkgoacae). Their maximum level in phytopharmaceuticals containing ginkgo extracts has been recently restricted to 5 microg/g by the Commission E of the former

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service