Skip to Content
Merck
All Photos(1)

Documents

55188

Supelco

Sodium (R)-β-hydroxyisobutyrate

analytical standard

Synonym(s):

(R)-β-Hydroxyisobutyric acid sodium salt, (R)-3-Hydroxy-2-methylpropionic acid sodium salt, R-HIBA, R-β-HIBA-Na, Sodium (R)-3-hydroxy-2-methylpropionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NaO3
CAS Number:
Molecular Weight:
126.09
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

Assay

≥96.0% (GC)

optical purity

enantiomeric excess: ≥98.0%

shelf life

limited shelf life, expiry date on the label

impurities

≤5.0% water (cal. from elemental analysis)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

[Na+].C[C@H](CO)C([O-])=O

InChI

1S/C4H8O3.Na/c1-3(2-5)4(6)7;/h3,5H,2H2,1H3,(H,6,7);/q;+1/p-1/t3-;/m1./s1

InChI key

RBJZIQZDAZLXEK-AENDTGMFSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service