Skip to Content
Merck
All Photos(1)

Documents

51465

Sigma-Aldrich

Urea

tested according to Ph. Eur.

Synonym(s):

Ureum, Carbamide, Carbonyldiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Agency

USP/NF
tested according to Ph. Eur.

Assay

98.5-101.5% (Dried Substance)

form

solid

mp

132-135 °C (lit.)

solubility

H2O: soluble 480 g/L at 20 °C (completely)

density

1.335 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A mechanically and electrically self-healing supercapacitor.
Hua Wang et al.
Advanced materials (Deerfield Beach, Fla.), 26(22), 3638-3643 (2014-02-21)
Oliver C Witard et al.
Brain, behavior, and immunity, 39, 211-219 (2013-10-15)
The present study examined whether a high protein diet prevents the impaired leukocyte redistribution in response to acute exercise caused by a large volume of high-intensity exercise training. Eight cyclists (VO2max: 64.2±6.5mLkg(-1)min(-1)) undertook two separate weeks of high-intensity training while
Andrii Slonchak et al.
Journal of virology, 88(15), 8457-8467 (2014-05-16)
West Nile virus (WNV) is an enveloped virus with a single-stranded positive-sense RNA genome from the Flaviviridae family. WNV is spread by mosquitoes and able to infect humans, causing encephalitis and meningitis that can be fatal; it therefore presents a
Marco Benevento et al.
Nature communications, 5, 5613-5613 (2014-12-11)
The ectopic expression of Oct4, Klf4, c-Myc and Sox2 (OKMS) transcription factors allows reprogramming of somatic cells into induced pluripotent stem cells (iPSCs). The reprogramming process, which involves a complex network of molecular events, is not yet fully characterized. Here
G L French et al.
Lancet (London, England), 2(8551), 117-119 (1987-07-18)
Tuberculostearic acid, a structural component of Mycobacterium tuberculosis, was identified by gas chromatography/mass spectrometry with selected ion monitoring in cerebrospinal fluid (CSF) from 13 patients with proven and 8 out of 9 patients with suspected tuberculous meningitis; the negative result

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service