Skip to Content
Merck
All Photos(2)

Documents

51208

Millipore

LB Broth

NutriSelect® Plus, High salt, powder, pack of 500 g or 2.5 kg, non-sterile, nonselective for Escherichia coli

Synonym(s):

LB-Broth, high salt, Luria Bertani Broth, high salt

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

casein enzymic hydrolysate, 10 g/L
High salt
sodium chloride, 10 g/L
yeast extract, 5 g/L

manufacturer/tradename

NutriSelect® Plus

packaging

pack of 500 g or 2.5 kg

technique(s)

microbiological culture: suitable

final pH

7.5±0.2 (25 °C)

application(s)

food and beverages
life science and biopharma
microbiology

suitability

nonselective for Escherichia coli (Recombinant)
nonselective for coliforms

Looking for similar products? Visit Product Comparison Guide

Application

Luria Media are used for the cultivation and maintenance of recombinant strains of E. coli for genetic and molecular studies.

Preparation Note

Suspend 25 g in 1000 ml of purified water. Heat with frequent agitation and boil for one minute. Sterilize at 121°C for 15 minutes. Dispense as desired.

Storage and Stability

  • Store prepared media below 8°C, protected from direct light.
  • Store dehydrated powder, in a dry place, in tightly-sealed containers at 2-25°C.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service