Skip to Content
Merck
All Photos(2)

Key Documents

510998

Sigma-Aldrich

Bromochlorophenol Blue

Dye content 95 %

Synonym(s):

3′,3″-Dibromo-5′,5″-dichlorophenolsulfonephthalein

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€200.00

€200.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€200.00

About This Item

Empirical Formula (Hill Notation):
C19H10Br2Cl2O5S
CAS Number:
Molecular Weight:
581.06
Beilstein:
365559
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€200.00


Estimated to ship on28 May 2025


Request a Bulk Order

form

powder

Quality Level

composition

Dye content, 95%

visual transition interval

3.0-4.6, yellow to purple

mp

230 °C (dec.) (lit.)

solubility

ethanol: 10 mg/mL, clear

λmax

590 nm

ε (extinction coefficient)

≥15000 at 306-310 nm in 0.1 M NaOH at 0.01 g/L
≥8000 at 378-382 nm in 0.1 M NaOH at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1c(Cl)cc(cc1Br)C2(OS(=O)(=O)c3ccccc23)c4cc(Cl)c(O)c(Br)c4

InChI

1S/C19H10Br2Cl2O5S/c20-12-5-9(7-14(22)17(12)24)19(10-6-13(21)18(25)15(23)8-10)11-3-1-2-4-16(11)29(26,27)28-19/h1-8,24-25H

InChI key

MDGFKZKMIQQRPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael D Green et al.
Emerging infectious diseases, 14(4), 552-556 (2008-04-09)
We developed a colorimetric and chromatographic assay for oseltamivir to assess the authenticity of Tamiflu (F. Hoffmann-La Roche Ltd., Basel, Switzerland) because of a growing concern about counterfeit oseltamivir. The colorimetric assay is quantitative and relies on an extractable colored
Feng Gao et al.
Combinatorial chemistry & high throughput screening, 9(5), 381-388 (2006-06-22)
Protein chip technology provides a new and useful tool for high-throughput screening of drugs because of its high performance and low sample consumption. In order to screen elastase inhibitors on a large scale, we designed a composite microarray integrating enzyme
Andrés H Thomas et al.
Biochimica et biophysica acta, 1858(1), 139-145 (2015-11-10)
Oxidized pterins, efficient photosensitizers under UVA irradiation, accumulate in the skin of patients suffering from vitiligo, a chronic depigmentation disorder. Soybean phosphatidylcholine (SoyPC) liposomes were employed as model membranes to investigate if pterin (Ptr), the parent compound of oxidized pterins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service