Skip to Content
Merck
All Photos(1)

Key Documents

48307

Supelco

Bile acids-selective liquid ion-exchanger microelectrode - Cocktail A

Selectophore

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700

product line

Selectophore

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Determination of bile acid activity in living liver tissue with a micro-electrode

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biotechnol. Progress, 6, 62-62 (1990)
B Yu et al.
Biosensors & bioelectronics, 5(3), 215-222 (1990-01-01)
The preparation of a compound liquid ion exchanger used for the miniaturization of bile acid sensitive microelectrode is described. The liquid ion exchanger is 2% benzyldimethylhexadecylammonium cholate in decanol/0.1 M hexadecyltributylammonium taurocholate in 5% hexachlorobenzene, 0.5% bromoacetanilide O-dichlorobenzene. The slopes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service