Skip to Content
Merck
All Photos(1)

Documents

46729

Supelco

Salinomycin monosodium salt

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H69NaO11
CAS Number:
Molecular Weight:
772.98
Beilstein:
4901827
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E716
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
clinical
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O[C@H]1[C@@]2(O[C@@](CC2)([C@]3([H])O[C@H]([C@](O)(CC3)CC)C)C)O[C@]4(C=C1)O[C@]([C@H](C[C@H]4C)C)([H])[C@@H](CC)C([C@@H](C)[C@@H](O)[C@@H]([C@]5([H])O[C@@](CC[C@@H]5C)([H])[C@@H](CC)C(O[Na])=O)C)=O

InChI

1S/C42H70O11.Na/c1-11-29(38(46)47)31-15-14-23(4)36(50-31)27(8)34(44)26(7)35(45)30(12-2)37-24(5)22-25(6)41(51-37)19-16-32(43)42(53-41)21-20-39(10,52-42)33-17-18-40(48,13-3)28(9)49-33;/h16,19,23-34,36-37,43-44,48H,11-15,17-18,20-22H2,1-10H3,(H,46,47);/q;+1/p-1/t23-,24-,25+,26-,27-,28-,29+,30-,31+,32+,33+,34+,36+,37-,39-,40+,41-,42-;/m0./s1

InChI key

YPZYGIQXBGHDBH-UZHRAPRISA-M

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Małgorzata Olejnik et al.
Rapid communications in mass spectrometry : RCM, 32(8), 629-634 (2018-02-15)
Salinomycin is an ionophore antibiotic with potential anticancer activity. The history of its use in veterinary medicine shows large differences in species susceptibility to its toxicity. At the same time, the results of research to date suggest a correlation between
Jaganmohan Reddy Jangamreddy et al.
Biochimica et biophysica acta, 1833(9), 2057-2069 (2013-05-04)
The molecular mechanism of Salinomycin's toxicity is not fully understood. Various studies reported that Ca(2+), cytochrome c, and caspase activation play a role in Salinomycin-induced cytotoxicity. Furthermore, Salinomycin may target Wnt/β-catenin signaling pathway to promote differentiation and thus elimination of
Lidia Radko et al.
Toxicology in vitro : an international journal published in association with BIBRA, 52, 314-320 (2018-07-18)
Salinomycin (SAL) is a polyether antibiotic, which is commonly used as a coccidiostat and has recently shown to exhibit anticancer activity. The toxic action of the drug may be connected with the extent and routes of its biotransformation. The cytotoxic
Jin Zhou et al.
Toxicology letters, 222(2), 139-145 (2013-08-07)
Postoperative chemotherapy for Colorectal cancer (CRC) patients is not all effective and the main reason might lie in cancer stem cells (CSCs). Emerging studies showed that CSCs overexpress some drug-resistance related proteins, which efficiently transport the chemotherapeutics out of cancer
Danxin Wu et al.
Biochemical and biophysical research communications, 443(2), 712-717 (2013-12-18)
Salinomycin (Sal) is a polyether ionophore antibiotic that has recently been shown to induce cell death in various human cancer cells. However, whether salinomycin plays a functional role in nasopharyngeal carcinoma (NPC) has not been determined to date. The present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service