Skip to Content
Merck
All Photos(1)

Key Documents

46391

Supelco

Dicyclanil

PESTANAL®, analytical standard

Synonym(s):

4,6-Diamino-2-(cyclopropylamino)-5-pyrimidinecarbonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€121.00

€121.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€121.00

About This Item

Empirical Formula (Hill Notation):
C8H10N6
CAS Number:
Molecular Weight:
190.21
Beilstein:
9265760
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€121.00


Estimated to ship on22 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1nc(NC2CC2)nc(N)c1C#N

InChI

1S/C8H10N6/c9-3-5-6(10)13-8(14-7(5)11)12-4-1-2-4/h4H,1-2H2,(H5,10,11,12,13,14)

InChI key

PKTIFYGCWCQRSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Dicyclanil is an insect growth regulator belonging to the group of pyrimidinamines.[1]

Application

Dicyclanil may be used as an analytical reference standard for the quantification of the analyte in biological samples[1], bovine, porcine and chicken muscle[2] using various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael K Rust et al.
Journal of medical entomology, 54(2), 418-421 (2016-12-25)
Insect growth regulators (IGRs) such as lufenuron, methoprene, and pyriproxyfen have been important tools in the integrated pest management of cat fleas, Ctenocephalides felis (Bouché), for the past two decades. Other IGRs have been registered for the control of termite
P J James et al.
Australian veterinary journal, 95(8), 265-272 (2017-07-28)
To determine the prophylactic and therapeutic efficacy of Australian-registered insecticide formulations against Old World screwworm (OWS) myiases for potential use in screwworm containment and eradication programs. The longevity of protection provided by six insecticidal formulations (subcutaneous ivermectin, doramectin and abamectin
Simultaneous determination of cyromazine and dicyclanil in animal edible tissues using UPLC-MS/MS
Hou X, et al.
Food Additives & Contaminants. Part A, Chemistry, Analysis, Control, Exposure & Risk Assessment, 30(4), 660-665 (2013)
Matthew J Traylor et al.
Insect biochemistry and molecular biology, 90, 14-22 (2017-09-18)
The Australian sheep blowfly, Lucilia cuprina, is a primary cause of sheep flystrike and a major agricultural pest. Cytochrome P450 enzymes have been implicated in the resistance of L. cuprina to several classes of insecticides. In particular, CYP6G3 is a
Lu-Qi Zhang et al.
Journal of mass spectrometry : JMS, 55(3), e4497-e4497 (2020-01-10)
A multiclass and multiresidue method for screening veterinary drugs and pesticides in infant formula was developed and validated using ultrahigh-performance liquid chromatography coupled to Quadrupole-Orbitrap high-resolution mass spectrometry (UHPLC-HRMS). A total of 49 veterinary drugs and pesticides investigated belong to

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service