Skip to Content
Merck
All Photos(1)

Documents

46207

Supelco

Diethylstilbestrol

VETRANAL®, analytical standard

Synonym(s):

(E)-3,4-Bis(4-hydroxyphenyl)-3-hexene, DES, Stilbestrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20O2
CAS Number:
Molecular Weight:
268.35
Beilstein:
2056095
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC\C(c1ccc(O)cc1)=C(\CC)c2ccc(O)cc2

InChI

1S/C18H20O2/c1-3-17(13-5-9-15(19)10-6-13)18(4-2)14-7-11-16(20)12-8-14/h5-12,19-20H,3-4H2,1-2H3/b18-17+

InChI key

RGLYKWWBQGJZGM-ISLYRVAYSA-N

Gene Information

human ... ESR1(2099)

Looking for similar products? Visit Product Comparison Guide

General description

Diethylstilbestrol (DES) belongs to the synthetic oestrogens class of compounds.

Application

DES has been used as a reference standard for the determination of DES in:
  • Dairy products by QuEChERS (quick, easy, cheap, effective, rugged, and safe) extraction procedure and ultra-high performance liquid chromatography-triple quadrupole mass spectrometry (UHPLC-QqQ MS/MS) equipped with electrospray ionization (ESI) and multiple reaction monitoring (MRM) mode of detection.
  • Water samples by magnetic micro-dispersive solid-phase extraction (m-μSPE) and HPLC-MS.

It may be used as a reference standard for the analysis of DES in
  • Aquatic products by modified QuEChERS extraction and multiplug filtration clean-up (m-PFC) followed by LC-ESI-MS/MS with MRM mode of detection.
  • Milk samples by vortex-assisted hollow fibre liquid-phase microextraction (VA-HF-LPME) and HPLC with diode array detection (HPLC-DAD).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Multiplug filtration cleanup method with multi-walled carbon nanotubes for the analysis of malachite green, diethylstilbestrol residues, and their metabolites in aquatic products by liquid chromatography-tandem mass spectrometry.
Qin Y, et al.
Analytical and Bioanalytical Chemistry, 408(21), 5801-5809 (2016)
Core-shell polydopamine magnetic nanoparticles as sorbent in micro-dispersive solid-phase extraction for the determination of estrogenic compounds in water samples prior to high-performance liquid chromatography-mass spectrometry analysis.
Socas-Rodriguez B, et al.
Journal of Chromatography A, 1397(7), 1-10 (2015)
Multiresidue analysis of oestrogenic compounds in cow, goat, sheep and human milk using core-shell polydopamine coated magnetic nanoparticles as extraction sorbent in micro-dispersive solid-phase extraction followed by ultra-high-performance liquid chromatography tandem mass spectrometry
Socas-Rodriguez B, et al.
Analytical and Bioanalytical Chemistry, 410(7), 2031-2042 (2018)
Vortex-assisted hollow fibre liquid-phase microextraction technique combined with high performance liquid chromatography-diode array detection for the determination of oestrogens in milk samples.
Wang P, et al.
Food Chemistry, 172(21), 385-390 (2015)
Multiresidue determination of estrogens in different dairy products by ultra-high-performance liquid chromatography triple quadrupole mass spectrometry.
Socas-Rodriguez B, et al.
Journal of Chromatography A, 1496(7), 58-67 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service