Skip to Content
Merck
All Photos(1)

Key Documents

45891

Supelco

Parathion-methyl solution

100 μg/mL in cyclohexane, PESTANAL®, analytical standard

Synonym(s):

Methyl parathion

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10NO5PS
CAS Number:
Molecular Weight:
263.21
Beilstein:
8905814
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in cyclohexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

S=P(OC)(OC)OC1=CC=C([N+]([O-])=O)C=C1

InChI

1S/C8H10NO5PS/c1-12-15(16,13-2)14-8-5-3-7(4-6-8)9(10)11/h3-6H,1-2H3

InChI key

RLBIQVVOMOPOHC-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Scott A Trammell et al.
Chemical communications (Cambridge, England), 48(34), 4121-4123 (2012-03-22)
Using a low power green laser, we have demonstrated a rate acceleration of ~2-fold for the hydrolysis of methyl parathion by irradiating the plasmon absorption band of Au nanoparticles capped with a Cu(bpy) catalyst.
Anirut Ekkhunnatham et al.
World journal of microbiology & biotechnology, 28(4), 1739-1746 (2012-07-19)
Methyl parathion hydrolase (MPH) from a methyl parathion-degrading Burkholderia cepacia indigenous to Thailand was purified to apparent homogeneity by three steps of column chromatography using Resource S, Sephadex G100, and Octyl Sepharose 4FF columns. Its molecular mass was determined to
Jingming Gong et al.
Biosensors & bioelectronics, 39(1), 320-323 (2012-08-08)
We developed a highly sensitive flow injection/amperometric biosensor for the detection of organophosphate pesticides (OPs) using layered double hydroxides (LDHs) as the immobilization matrix of acetylcholinesterase (AChE). LDHs provided a biocompatible microenvironment to keep the bioactivity of AChE, due to
Xiang Huang et al.
Aquatic toxicology (Amsterdam, Netherlands), 114-115, 189-199 (2012-03-27)
Methyl parathion (MP) is a widely used organophosphorus pesticide that causes severe health and environmental effects. We investigated the alteration of the proteomic profile in the membrane enriched fraction of the kidneys of the scallop Mizuhopecten yessoensis exposed to low-level
Thiago M R Araújo et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 48(4), 251-259 (2013-02-05)
This work presents a study of the abiotic degradation of commercially available methyl parathion in aqueous solution at two different concentrations (88 mg/L and 200 μg/L). The effects of solar irradiation and the presence of humic acids were evaluated and

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service