Skip to Content
Merck
All Photos(1)

Key Documents

45854

Supelco

Fenitrothion solution

100 ng/μL in cyclohexane, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12NO5PS
CAS Number:
Molecular Weight:
277.23
Beilstein:
8983553
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 ng/μL in cyclohexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

COP(=S)(OC)Oc1ccc(c(C)c1)[N+]([O-])=O

InChI

1S/C9H12NO5PS/c1-7-6-8(4-5-9(7)10(11)12)15-16(17,13-2)14-3/h4-6H,1-3H3

InChI key

ZNOLGFHPUIJIMJ-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-0.4 °F

Flash Point(C)

-18 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yihui Luo et al.
Analytical biochemistry, 421(1), 130-137 (2011-12-06)
A single-chain variable fragment (ScFv) complementary DNA (cDNA) library against fenitrothion was constructed, and ScFvs specific for fenitrothion were selected by ribosome display from the library. After three rounds of ribosome display, the ScFv genes were cloned into Escherichia coli
Jong Sung Lim et al.
Journal of bacteriology, 194(4), 896-896 (2012-01-26)
Burkholderia species are ubiquitous in soil environments. Many Burkholderia species isolated from various environments have the potential to biodegrade man-made chemicals. Burkholderia sp. strain YI23 was isolated from a golf course soil and identified as a fenitrothion-degrading bacterium. In this
Muhammet Mustafa Akıner et al.
Turkiye parazitolojii dergisi, 36(2), 87-91 (2012-07-18)
The objective of this study is to assess the resistance status and yearly changes involved for house fly populations from six cities in Turkey. Field strains of house fly (Musca domestica L. Diptera: Muscidae) were collected in 2004-2006 from cow
Fatma M El-Demerdash
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 47(4), 262-268 (2012-03-21)
A mixture of pyrethroids plus organophosphates was assessed for their potential effects on lipid peroxidation, the antioxidant defense system and lactate dehydrogenase (LDH) in rat kidney in vitro. Various insecticide concentrations were incubated with kidney homogenate at 37°C for different
H Ikeura et al.
Journal of food science, 78(2), T350-T355 (2013-01-15)
In this study, the effects of ozone microbubbles (OMCB) treatment at various water temperatures on the removal of residual fenitrothion (FT) pesticides and on the physical properties of lettuce and cherry tomatoes was examined. The residual FT percentage in lettuce

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service