Skip to Content
Merck
All Photos(1)

Key Documents

45778

Supelco

1,2-Dihydronaphthalene

analytical standard

Synonym(s):

Dialin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10
CAS Number:
Molecular Weight:
130.19
Beilstein:
1851372
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.582 (lit.)

bp

89 °C/16 mmHg (lit.)

mp

−8 °C (lit.)

density

0.997 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

C1Cc2ccccc2C=C1

InChI

1S/C10H10/c1-2-6-10-8-4-3-7-9(10)5-1/h1-3,5-7H,4,8H2

InChI key

KEIFWROAQVVDBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dihydronaphthalene is a bicyclic hydrocarbon, which resembles naphthalene but shows partial unsaturation in one of its rings. Its derivatives find wide applications in natural compounds of therapeutic interest.

Application

1,2-Dihydronaphthalene may be used as an analytical standard for the determination of the analyte in polycyclic aromatic hydrocarbon (PAH) mixtures by gas chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Retention indices for programmed-temperature capillary-column gas chromatography of polycyclic aromatic hydrocarbons
Lee ML, et al.
Analytical Chemistry, 51(6), 768-773 (1979)
Prediction of gas chromatographic retention indexes of polycyclic aromation compounds and nitrated polycyclic aromatic compounds
Rohrbaugh RH and Jurs PC
Analytical Chemistry, 58(6), 1210-1212 (1986)
The synthesis of novel dihydronaphthalenes and benzofluorenes
Novel Selenium-Mediated Rearrangements and Cyclisations, 77(6), 1210-1212 (2012)
Márcia Kameyama et al.
Molecules (Basel, Switzerland), 16(11), 9421-9438 (2011-11-15)
A new approach for the synthesis of indatraline was developed using as the key step an iodine(III)-mediated ring contraction of a 1,2-dihydronaphthalene derivative. Behavioral tests were conducted to evaluate the effect of indatraline and of its precursor indanamide on the
Claudia Sanfilippo et al.
Biotechnology letters, 26(23), 1815-1819 (2005-01-27)
Chloroperoxidase from Caldariomyces fumago catalyses the oxidation of 1,2-dihydronaphthalene to (1R,2R)-(+)-dihydroxytetrahydronaphthalene in homogenous citrate buffer/ionic liquid mixtures, using t-butyl hydroperoxide as O2 donor. It tolerates up to 30 (v/v) 1,3-dimethylimidazolium methylsulfate or 1-butyl-3-methylimidazolium methylsulfate. The enzyme activity in these ionic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service