Skip to Content
Merck
All Photos(1)

Key Documents

45465

Supelco

Dodemorph

PESTANAL®, analytical standard

Synonym(s):

4-Cyclododecyl-2,6-dimethylmorpholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H35NO
CAS Number:
Molecular Weight:
281.48
Beilstein:
511312
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

description

cis-trans mixture of isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1CN(CC(C)O1)C2CCCCCCCCCCC2

InChI

1S/C18H35NO/c1-16-14-19(15-17(2)20-16)18-12-10-8-6-4-3-5-7-9-11-13-18/h16-18H,3-15H2,1-2H3

InChI key

JMXKCYUTURMERF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Skin Corr. 1C - Skin Sens. 1A - STOT RE 2

Target Organs

Liver

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L H Leenheers et al.
Journal of chromatographic science, 30(6), 228-232 (1992-06-01)
As part of a health hazard survey of occupational exposure to pesticides in greenhouse growing of ornamentals, analytical methods are developed and validated for measurement of exposure of workers to the fungicide dodemorph. A gas chromatographic method is developed using
Katrin Vorkamp et al.
Chemosphere, 53(5), 505-514 (2003-09-02)
The aim of this study was to examine the stability and distribution of the fungicide dodemorph during anaerobic digestion of biological waste, including a short subsequent aerobic treatment. Three reactor experiments were conducted. The influences of pH and dissolved organic
R Kreuzig et al.
Journal of biochemical and biophysical methods, 43(1-3), 403-409 (2000-06-28)
The applicability of supercritical fluid extraction (SFE) in pesticide residue analysis in soil was investigated by analysing real soil samples from field experiments. Additionally, radiotracer batch experiments were performed to study the release of non-extractable residues. High repeatability, accuracy and
R Brouwer et al.
Archives of environmental contamination and toxicology, 23(3), 273-280 (1992-10-01)
On 18 farms for rose culture in greenhouses in The Netherlands, dermal exposure of hands and forearms to abamectin (avermectin B1), dodemorph (4-cyclododecyl-2,6-dimethylmorpholinium acetate) and bupirimate (5-butyl-2-(ethylamino)-6-methyl-4-pyrimidinyl dimethylsulphate) was measured during crop activities. Dermal exposure during cutting (75 workers) amounted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service