Skip to Content
Merck
All Photos(1)

Key Documents

45443

Supelco

Dienochlor

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10Cl10
CAS Number:
Molecular Weight:
474.64
Beilstein:
2064747
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

ClC1=C(Cl)C(Cl)(C(Cl)=C1Cl)C2(Cl)C(Cl)=C(Cl)C(Cl)=C2Cl

InChI

1S/C10Cl10/c11-1-2(12)6(16)9(19,5(1)15)10(20)7(17)3(13)4(14)8(10)18

InChI key

LWLJUMBEZJHXHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Fruetel et al.
Chemical research in toxicology, 7(4), 487-494 (1994-07-01)
Dienochlor (Pentac) (C10Cl10) has been used for 30 years as a miticide with little knowledge of its mode of action or metabolic fate except that it is quickly degraded by rats. This study examines the reactions of dienochlor with GSH
G B Quistad et al.
Toxicology and applied pharmacology, 85(2), 215-220 (1986-09-15)
Within four days of receiving a single oral dose (1 mg/kg) of [U-ring-14C]dienochlor [bis(pentachloro-2,4-cyclopentadien-1-yl)] female rats excreted 2 and 88% of the applied 14C in urine and feces, respectively. Metabolites could not be identified and the preponderance of the fecal
D Skandrani et al.
Toxicology in vitro : an international journal published in association with BIBRA, 20(8), 1378-1386 (2006-08-04)
Two organochlorines (dienochlor, endosulfan) and one neonicotinoid (imidacloprid) insecticides were investigated as putative cellular aggressors, both as pure chemicals and as commercial formulations, in order to evaluate the additional toxicity due to additives present in the commercial formulations. Toxicity was
Survey of pesticide use by Ontario greenhouse chrysanthemum producers.
B A Archibald et al.
Bulletin of environmental contamination and toxicology, 53(4), 486-492 (1994-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service