Skip to Content
Merck
All Photos(1)

Key Documents

44156

Supelco

N-Methyl-N-trimethylsilyltrifluoroacetamide activated II

for GC derivatization, LiChropur, activated with 2-(Trimethylsilyl)ethanethiol

Synonym(s):

MSTFA activated II

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116105
NACRES:
NA.22
Pricing and availability is not currently available.

grade

derivatization grade ((GC derivatization))
for GC derivatization

Quality Level

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.380

storage temp.

2-8°C

Application

  • Derivatizing agent for the trimethylsilylation of -OH and -NH groups for GC and GC-MS[1]
  • Suitable for the silylation of testosterone, tested by GCMS

Features and Benefits

  • Activated with trimethylsily-ethanethiol.       
  • This mixture is equivalent to a 1000:2 mixture of MSTFA and iodotrimethylsilane.       

Other Notes

This mixture is equivalent to a 1000 : 2 mixture of MSTFA and iodotrimethylsilane.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F

Flash Point(C)

26 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W. Schanzer et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 687, 93-93 (1996)

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service