Skip to Content
Merck

Skip To

43301

Enalaprilat

analytical standard

Synonym(s):

(2S)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenyl-propyl]amino]propanoyl]pyrrolidine-2-carboxylic acid, N-[(1S)-1-Carboxy-3-phenylpropyl]-L-alanyl-L-proline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 124,00

About This Item

Empirical Formula (Hill Notation):
C18H24N2O5
Molecular Weight:
348.39
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

€ 124,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

impurities

5-12 wt. % water

format

neat

SMILES string

O=C(N1CCC[C@H]1C(O)=O)[C@H](C)N[C@H](C(O)=O)CCC2=CC=CC=C2

InChI

1S/C18H24N2O5/c1-12(16(21)20-11-5-8-15(20)18(24)25)19-14(17(22)23)10-9-13-6-3-2-4-7-13/h2-4,6-7,12,14-15,19H,5,8-11H2,1H3,(H,22,23)(H,24,25)/t12-,14-,15-/m0/s1

InChI key

LZFZMUMEGBBDTC-QEJZJMRPSA-N

Gene Information

human ... ACE(1636)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1235274PHR3440Y0000615
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

-

grade

analytical standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

assay

≥98.0% (HPLC)

assay

-

assay

-

assay

-

format

neat

format

neat

format

-

format

neat

Quality Level

100

Quality Level

-

Quality Level

300

Quality Level

-

Gene Information

human ... ACE(1636)

Gene Information

human ... ACE(1636)

Gene Information

-

Gene Information

human ... ACE(1636)


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters