43236
2-Ethyl-3,5(6)-dimethylpyrazine, mixture of isomers
analytical standard
Synonym(s):
2-Ethyl-3(5 or 6)-dimethylpyrazine, mixture of isomers
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥98.0% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
impurities
≤2.0% water
refractive index
n20/D 1.500-1.502
n20/D 1.5015 (lit.)
bp
180-181 °C (lit.)
density
0.965 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
neat
storage temp.
2-8°C
SMILES string
CCc1ncc(C)nc1C.CCc2nc(C)cnc2C
InChI
1S/2C8H12N2/c1-4-8-7(3)10-6(2)5-9-8;1-4-8-7(3)9-5-6(2)10-8/h2*5H,4H2,1-3H3
InChI key
BOFLOMVCGPWPQC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
156.2 °F
Flash Point(C)
69 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service