Skip to Content
Merck
All Photos(1)

Key Documents

428787

Supelco

Amberlite HPR2900 Ion Exchange Resin

hydrogen form, 25-35 mesh

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€125.00
2.5 KG
€668.00

€125.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€125.00
2.5 KG
€668.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

€125.00


Check Cart for Availability

Request a Bulk Order

Product Name

AmberLite HPR2900 H hydrogen form, hydrogen form

description

formerly Dowex Marathon MSC H

Quality Level

form

beads

crosslinking

>95 % cross-linked

parameter

150 °C max. temp.

moisture

50-56%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (macroreticular)

matrix active group

sulfonic acid

particle size

24-29 mesh
525-625 μm

operating pH

0-14

capacity

1.6 meq/mL by wetted bed volume

separation technique

cation exchange

SMILES string

C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The Dowex Marathon MSC is a superior resin to the Dowex MSC-1. The Dowex Marathon series provides narrower particle distribution, better efficiency, and improved kinetics when compared to the gaussian Dowex products.

Application

Dowex Marathon MSC hydrogen form was used to remove active copper catalysts remaining in redissolved solution.[1]
Strongly acidic cation exchange resin for water demineralization. This resin meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Features and Benefits

Strong cation

Other Notes

This ion-exchange resin has not been specially processed nor cleaned. We suggest that it be treated to a suitable preliminary elution and wash.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
Marathon is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sharon Y Wong et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 17(3), 480-490 (2009-01-15)
Three combinatorial libraries of polymeric vectors were evaluated to investigate the functional roles of molecular weight (MW), cations, pH-sensitive moieties, and hydrophobic derivitization in polymer-mediated gene delivery. Four cationic and pH-sensitive moieties (imidazole, primary, secondary, and tertiary amino) and three

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. How can I convert a cation exchange resin from the hydrogen form to the sodium form?

    1 answer
    1. To convert a cation exhcange resin from the hydrogen form to the sodium form, use two to four bed volumes of 0.5-2.0% NaCl or NaOH.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service