Skip to Content
Merck
All Photos(1)

Key Documents

40858

Supelco

Propyl acetate

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€123.00

€123.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€123.00

About This Item

Linear Formula:
CH3COOCH2CH2CH3
CAS Number:
Molecular Weight:
102.13
Beilstein:
1740764
EC Number:
MDL number:
UNSPSC Code:
85151701
eCl@ss:
39022103
PubChem Substance ID:
NACRES:
NA.24

€123.00


Estimated to ship on02 June 2025


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

3.5 (vs air)

vapor pressure

25 mmHg ( 20 °C)

Assay

≥99.7% (GC)

autoignition temp.

842 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

1.7 %, 37 °F
8 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.384 (lit.)
n20/D 1.384

bp

102 °C (lit.)

mp

−95 °C (lit.)

solubility

water: soluble

density

0.888 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCOC(C)=O

InChI

1S/C5H10O2/c1-3-4-7-5(2)6/h3-4H2,1-2H3

InChI key

YKYONYBAUNKHLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Propyl acetate may be used in the preparation of geranyl acetate.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.2 °F - closed cup

Flash Point(C)

11.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saturated vapour pressure of methyl acetate, ethyl acetate, n-propyl acetate, methyl propionate, and ethyl propionate.
Polak J and Mertl I
Collection of Czechoslovak Chemical Communications, 30, 3526-3528 (1965)
Excess molar enthalpies for binary mixtures of n-propanol, acetic acid, and n-propyl acetate at 313.15 K and atmospheric pressure.
Letyanina I, et al.
Fluid Phase Equilibria, 381, 77-82 (2014)
Kinetics study of propyl acetate synthesis reaction catalyzed by Amberlyst 15.
Huang YS and Sundmacher K.
International Journal of Chemical Kinetics, 39(5), 245-253 (2007)
Geranyl acetate synthesis by lipase-catalyzed transesterification in supercritical carbon dioxide.
Geranyl acetate synthesis by lipase-catalyzed transesterification in supercritical carbon dioxide.
Chulalaksananukul W, et al.
Enzyme and Microbial Technology, 15(8), 691-698 (1993)
Lan-Chun Nie et al.
Zhi wu sheng li yu fen zi sheng wu xue xue bao = Journal of plant physiology and molecular biology, 31(6), 663-667 (2005-12-20)
Aroma volatiles from apple (Malus domestica Borkh. var. Starkrimson) fruit at different stages of maturity were collected by solid adsorbent-Tenax-GC and determined by thermodesorption and GC-MS. Production of propyl acetate, butyl acetate, ethyl 2-methyl-butanoate and total ester volatiles and changes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service