40252
N,N-Dimethylformamide diethyl acetal
for esterification of fatty acids, ≥95.0% (GC)
Synonym(s):
1,1-Diethoxy-N,N-dimethylmethylamine, 1,1-Diethoxytrimethylamine
About This Item
Recommended Products
Quality Level
Assay
≥95.0% (GC)
quality
for esterification of fatty acids
color
colorless to yellow
refractive index
n20/D 1.397-1.401
n20/D 1.400 (lit.)
bp
130-133 °C (lit.)
density
0.859 g/mL at 25 °C (lit.)
SMILES string
CCOC(OCC)N(C)C
InChI
1S/C7H17NO2/c1-5-9-7(8(3)4)10-6-2/h7H,5-6H2,1-4H3
InChI key
BWKAYBPLDRWMCJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
82.4 °F - closed cup
Flash Point(C)
28 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service