Skip to Content
Merck
All Photos(1)

Key Documents

39363

Supelco

Sodium 3-(3,4-dihydroxyphenyl)-DL-lactate

analytical standard

Synonym(s):

α,3,4-Trihydroxybenzenepropanoic acid sodium salt, 3-(3,4-Dihydroxyphenyl)lactic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NaO5
CAS Number:
Molecular Weight:
220.15
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤12.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

[Na+].OC(Cc1ccc(O)c(O)c1)C([O-])=O

InChI

1S/C9H10O5.Na/c10-6-2-1-5(3-7(6)11)4-8(12)9(13)14;/h1-3,8,10-12H,4H2,(H,13,14);/q;+1/p-1

InChI key

ZMMKVDBZTXUHFO-UHFFFAOYSA-M

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Juan Li et al.
Clinical hemorheology and microcirculation, 50(4), 267-278 (2012-01-14)
This study intended to examine the effect of 3,4-dihydroxyphenyl lactic acid (DLA), a major ingredient of Salvia miltiorrhiza, on lipopolysaccahride (LPS) -induced mouse cerebral cortical microcirculatory disturbance. Velocity of red blood cells in, and albumin leakage from venules, and the
Yating Chong et al.
Molecular medicine reports, 5(3), 812-816 (2011-12-03)
P-glycoprotein (P-gp) plays a key role in the distribution and elimination of many clinically important therapeutic substances. P-gp inhibition is capable of increasing the concentration of various drugs in the brain. Danshensu is a main hydrophilic constituent of Salvia miltiorrhiza
Ying Yin et al.
European journal of pharmacology, 699(1-3), 219-226 (2012-12-04)
Danshensu, as the effective component of Salvia miltiorrhiza (Danshen), has been widely used in clinic for treatment of cardiovascular diseases in China. In the present study, we aimed to confirm the cardioprotective effect of Danshensu from myocardial ischemia/reperfusion (MI/R) injury
Yaoling Jia et al.
European journal of medicinal chemistry, 55, 176-187 (2012-07-31)
A series of novel amide and thioester conjugates between Danshensu and cysteine derivatives have been designed and synthesized based on the strategy of "medicinal chemical hybridization". Pharmacological evaluation indicated that the amide conjugates 3a/4a/17a and thioester conjugates 6a-d exhibited obvious
Libo Du et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(4), 1281-1287 (2012-12-12)
The enhanced antioxidant activity of surface-functionalized gold nanoparticles (AuNPs) synthesized by self-assembly has attracted great attention, but little is known about the mechanism behind the enhanced activity. To address this challenge, the antioxidant activity of Au@PEG3SA (i.e., surface-functionalization of spherical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service