Skip to Content
Merck

39145

N-(7-Dimethylamino-4-methyl-3-coumarinyl)maleimide

≥99.0% (HPLC)

Synonym(s):

DACM-3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H14N2O4
CAS Number:
Molecular Weight:
298.29
EC Number:
259-499-0
UNSPSC Code:
12352200
PubChem Substance ID:
Beilstein/REAXYS Number:
1545346
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99.0% (HPLC)

mp

218-220 °C (lit.)

fluorescence

λex 398 nm; λem 482 nm in 0.1 M phosphate pH 7.0 (after derivatization with 2-mercaptoethanol)

suitability

suitable for fluorescence

SMILES string

CN(C)c1ccc2C(C)=C(N3C(=O)C=CC3=O)C(=O)Oc2c1

InChI

1S/C16H14N2O4/c1-9-11-5-4-10(17(2)3)8-12(11)22-16(21)15(9)18-13(19)6-7-14(18)20/h4-8H,1-3H3

InChI key

ADEORFBTPGKHRP-UHFFFAOYSA-N

Other Notes

Fluorescent reagent with high selectivity for protein thiols and with superior fluorescence and water solubility properties than dansyl chloride




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library