Skip to Content
Merck
All Photos(1)

Key Documents

38954

Millipore

Malt Extract Agar modified, Vegitone

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Vegitone Malt Extract Agar modified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.00 g/L
dextrin, 2.75 g/L
maltose, 12.75 g/L
peptone (vegetable), 0.78 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

4.7±0.2 (25 °C)

application(s)

agriculture
bioburden testing
clinical testing
environmental
food and beverages
pharmaceutical

microbiology

suitability

selective by low pH for Aspergillus spp.
selective by low pH for Candida spp.
selective by low pH for Penicillium spp.
selective by low pH for Pichia spp.
selective by low pH for Saccharomyces spp.
selective by low pH for Zygosaccharomyces spp.
selective by low pH for molds (General Media)
selective by low pH for yeasts (General Media)

Application

This medium is recommended for isolation, detection and enumeration of yeasts and molds.

Preparation Note

Suspend 31.28 g in 1000 ml distilled water. Add 2.35 g glycerol (Cat. No. 49767). Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. AVOID OVERHEATING.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inês Mendes et al.
PloS one, 8(7), e66523-e66523 (2013-07-23)
Saccharomyces cerevisiae strains from diverse natural habitats harbour a vast amount of phenotypic diversity, driven by interactions between yeast and the respective environment. In grape juice fermentations, strains are exposed to a wide array of biotic and abiotic stressors, which

Articles

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service