Skip to Content
Merck
All Photos(1)

Key Documents

38911

Sigma-Aldrich

Dimethyl ether

puriss., ≥99.9% (GC)

Synonym(s):

Methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2O
CAS Number:
Molecular Weight:
46.07
Beilstein:
1730743
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:

Pricing and availability is not currently available.

vapor density

1.62 (vs air)

vapor pressure

>760 mmHg ( 25 °C)

grade

puriss.

Assay

≥99.9% (GC)

autoignition temp.

662 °F

expl. lim.

27 %

bp

−24.8 °C (lit.)

mp

−141 °C (lit.)

SMILES string

COC

InChI

1S/C2H6O/c1-3-2/h1-2H3

Looking for similar products? Visit Product Comparison Guide

Packaging

pressure tin with 250 mL (net ~150 g)

Other Notes

Sales restrictions may apply

control valve

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

-41.8 °F - closed cup

Flash Point(C)

-41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruce L Yoder et al.
The Journal of chemical physics, 138(4), 044202-044202 (2013-02-08)
We present a new experimental configuration for the study of size-dependent, angle-resolved photoelectron and photoion spectra of weakly bound ultrafine aerosol particles targeted at particle sizes below ~20 nm. It combines single photon ionization by a tunable, table-top vacuum ultraviolet
Salma Parveen et al.
The journal of physical chemistry. A, 113(21), 6182-6191 (2009-05-09)
Theoretical calculations have been carried out using ab initio MP2 and B3LYP density functional methods to investigate the interaction between fluorinated dimethyl ethers (nF = 1-5) and water. Depending on the number of F atoms implanted on the dimethyl ethers
Xiao Liang et al.
The journal of physical chemistry. B, 115(25), 8199-8206 (2011-06-10)
The acid-catalyzed hydrolysis of dimethyl ether (DME) to methanol was examined using ab initio density functional metadynamics simulations. Diffusion of the acid proton from the aqueous medium, leading to the formation of a protonated DME, is 12.3 kcal mol(-1) activated
Kazuyuki Oshita et al.
Chemosphere, 78(9), 1148-1154 (2010-01-02)
We investigated whether polychlorinated biphenyls (PCBs) and water could be simultaneously removed from river sediment by solvent extraction using liquefied dimethyl ether (DME) as the extractant. DME exists in a gaseous state at normal temperature and pressure and can dissolve
M Naqvi et al.
Bioresource technology, 101(3), 937-944 (2009-09-22)
There is great interest in developing black liquor gasification technology over recent years for efficient recovery of bio-based residues in chemical pulp mills. Two potential technologies of producing dimethyl ether (DME) and methane (CH(4)) as alternative fuels from black liquor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service