Skip to Content
Merck
All Photos(1)

Key Documents

38116

Supelco

Dibenzyl phthalate

analytical standard

Synonym(s):

1,2-Benzenedicarboxylic acid 1,2-bis(methylphenyl) ester, NSC 4057, Phthalic acid dibenzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18O4
CAS Number:
Molecular Weight:
346.38
Beilstein:
2338250
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

application(s)

environmental

format

neat

SMILES string

O=C(OCC1=CC=CC=C1)C2=CC=CC=C2C(OCC3=CC=CC=C3)=O

InChI

1S/C22H18O4/c23-21(25-15-17-9-3-1-4-10-17)19-13-7-8-14-20(19)22(24)26-16-18-11-5-2-6-12-18/h1-14H,15-16H2

InChI key

UCVPKAZCQPRWAY-UHFFFAOYSA-N

General description

Dibenzyl phthalate, a diaryl phthalate, is extensively used as a plasticizer to modify the properties of the synthetic resin substrates, resulting in the improvement of flexibility and durability of the end products. It is considered as a potential environmental pollutant.

Application

Dibenzyl phthalate may be used as an analytical standard for the determination of the analyte in environmental samples, and benzyl butyl phthalate plasticizers used in lacquers by various chromatography techniques.

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Szycher's Handbook of Polyurethanes, 1174(1-2), 40-49 (1999)
Encyclopedic Dictionary of Polymers, Volume 1
Encyclopedic Dictionary of Polymers, (1-2), 40-49 (2010)
Development of a stir bar sorptive extraction and thermal desorption-gas chromatography-mass spectrometry method for the simultaneous determination of several persistent organic pollutants in water samples
Prieto A, et al.
Journal of Chromatography A, 1174(1-2), 40-49 (2007)
Identification and Determination of Plasticizers in Lacquers by Programmed Temperature Gas Chromatography.
Esposito GG
Analytical Chemistry, 35(10), 1439-1441 (1963)
Gas Chromatographic Analysis of High Boiling Point Plasticizers Using a Short Column.
Cook CD, et al.
Analytical Chemistry, 34(9), 1177-1178 (1962)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service