Skip to Content
Merck
All Photos(1)

Key Documents

37033

Supelco

Seneciphylline

analytical standard

Synonym(s):

α-Longilobin, 3,19-Didehydro-12-hydroxysenecionan-11,16-dione, Jacodine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23NO5
CAS Number:
Molecular Weight:
333.38
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

C\C=C1\CC(=C)[C@@](C)(O)C(=O)OCC2=CCN3CC[C@@H](OC1=O)[C@@H]23

InChI

1S/C18H23NO5/c1-4-12-9-11(2)18(3,22)17(21)23-10-13-5-7-19-8-6-14(15(13)19)24-16(12)20/h4-5,14-15,22H,2,6-10H2,1,3H3/b12-4-/t14-,15-,18-/m1/s1

InChI key

FCEVNJIUIMLVML-QPSVUOIXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Reconstitution

Reconstitution: Dried down, 50 μg/mL after reconstitution with 1 mL of water

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C C Yan et al.
Toxicology and applied pharmacology, 133(2), 277-284 (1995-08-01)
Despite their similarity in structure, pyrrolizidine alkaloids (PAs) vary in their LD50s and in the organs in which toxicity is expressed. We have examined whether there are differences in the metabolism of certain PAs that are associated with these quantitative
M Noorwala et al.
Fitoterapia, 71(5), 618-620 (2001-07-14)
The isolation and NMR spectra of otosenine (1) and seneciphylline (2) from Senecio lorenthii are reported.
H K Kakrani et al.
Journal of ethnopharmacology, 12(3), 271-278 (1984-12-01)
The effect of oral administration of the pyrrolizidine alkaloids, seneciphylline and senecionine, from Senecio vulgaris (Compositae) on activities of hepatic epoxide hydrase, glutathione-S-transferase, aminopyrine-N-demethylase and arylhydrocarbon hydroxylase (AHH) was investigated in microsomes of young male albino rats. Seneciphylline significantly increased
D F Eastman et al.
Drug metabolism and disposition: the biological fate of chemicals, 10(3), 236-240 (1982-05-01)
Two macrocyclic 14C-pyrrolizidine alkaloids (PA's), senecionine an seneciphylline, were studied regarding the distribution, excretion, transfer into milk, and covalent binding to hepatic macromolecules in BALB/c mice. After injection, radioactivity was rapidly excreted in the urine and feces (84% or greater)
Pyrrolizidine alkaloids from Melampyrum pratense.
E Roeder et al.
Natural toxins, 1(1), 35-37 (1992-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service