Skip to Content
Merck
All Photos(1)

Documents

36918

Supelco

2,4,6-Tribromophenol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br3C6H2OH
CAS Number:
Molecular Weight:
330.80
Beilstein:
776920
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

vapor density

11.4 (vs air)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

282-290 °C/746 mmHg (lit.)

mp

90-94 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Oc1c(Br)cc(Br)cc1Br

InChI

1S/C6H3Br3O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10H

InChI key

BSWWXRFVMJHFBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Pizarro et al.
Journal of chromatography. A, 1217(49), 7630-7637 (2010-11-03)
A dispersive liquid-liquid microextraction (DLLME) method has been optimised for simultaneously extracting 2,4,6-trichloranisole (TCA), 2,3,4,6-tetrachloroanisole (TeCA), 2,4,6-tribromoanisole (TBA), pentachloroanisole (PCA), 2,4,6-trichlorophenol (TCP), 2,3,4,6-tetrachlorophenol (TeCP), 2,4,6-tribromophenol (TBP) and pentachlorophenol (PCP) from wine. The haloanisoles and halophenols were automatically determined using a
Anna Norman Haldén et al.
Aquatic toxicology (Amsterdam, Netherlands), 100(1), 30-37 (2010-08-20)
The bromophenol 2,4,6-tribromophenol (TBP) is widely used as an industrial chemical, formed by degradation of tetrabromobisphenol-A, and it occurs naturally in marine organisms. Concentrations of TBP in fish have been related to intake via feed, but little is known about
Kristina Arnoldsson et al.
Environmental science & technology, 46(13), 7239-7244 (2012-06-13)
Polybrominated dibenzo-p-dioxins (PBDD) are emerging environmental pollutants with structural similarities to the highly characterized toxicants polychlorinated dibenzo-p-dioxins. The geographical and temporal variations of PBDD in biota samples from the Baltic Sea do not display features that are normally related to
K Y Kim et al.
Phytochemistry, 69(16), 2820-2825 (2008-10-28)
Diabetes mellitus is a most serious and chronic disease whose incidence rates are increasing with incidences of obesity and aging of the general population over the world. One therapeutic approach for decreasing postprandial hyperglycemia is to retard absorption of glucose
Natsuko Kajiwara et al.
Journal of hazardous materials, 192(3), 1250-1259 (2011-07-26)
The concentrations of traditional brominated flame retardants (BFRs) and organophosphate flame retardants (OPFRs) in new consumer products, including electronic equipment, curtains, wallpaper, and building materials, on the Japanese market in 2008 were investigated. Although some components of the electronic equipment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service