Skip to Content
Merck
All Photos(1)

Key Documents

36753

Supelco

PCB No 4

analytical standard

Synonym(s):

2,2′-Dichlorobiphenyl, 2,2′-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl2
CAS Number:
Molecular Weight:
223.10
Beilstein:
974158
Ballschmiter Number:
4
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

Clc1ccccc1-c2ccccc2Cl

InChI

1S/C12H8Cl2/c13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)14/h1-8H

InChI key

JAYCNKDKIKZTAF-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daesuk Chung et al.
Biology of reproduction, 73(5), 974-982 (2005-07-08)
The present study examined the hypothesis that inhibition of myometrial gap junctions through MAPK1-induced phosphorylation of GJA1 (connexin43) leads to inhibition of spontaneous phasic uterine contractions by 2,2'-dichlorobiphenyl (2,2'-DCB). Uterine strips from Gestation Day 10-pregnant rats exposed in muscle baths
M Wilbrink et al.
Xenobiotica; the fate of foreign compounds in biological systems, 21(2), 223-233 (1991-02-01)
1. The occurrence of an as yet unidentified cytochrome P-450 in the microsomal fraction of the digestive gland of the snail, Lymnaea stagnalis was studied. 2. Studies in vivo and in vitro (digestive gland homogenates or the 170,000g fraction) of
J C Bemis et al.
Neurotoxicology, 21(6), 1123-1134 (2001-03-10)
Assessments of the effects of exposure of human populations to complex environmental contaminants, such as those found in contaminated fish, necessitate the investigation of contaminant interactions. We have recently demonstrated that polychlorinated biphenyls (PCBs) and methylmercury (MeHg) synergistically reduce rat
W E Maier et al.
Journal of applied toxicology : JAT, 14(3), 225-229 (1994-05-01)
Polychlorinated biphenyl (PCBs) mixtures contain a number of different congeners, some of which have been proposed to be neuroactive. Recent studies have suggested that ortho-substituted PCBs may be neuroactive, while 'dioxin-like' non-ortho-substituted congeners are not. This study compared the in
O A Voie et al.
Archives of toxicology, 73(10-11), 588-593 (2000-02-09)
In this paper the effect of polychlorinated biphenyls (PCBs) on the production of reactive oxygen species (ROS) in rat synaptosomes is elucidated. The effect of methylmercury (MeHg) on rat synaptosomes was included as a positive control since several studies have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service