Skip to Content
Merck
All Photos(1)

Key Documents

36691

Sigma-Aldrich

Dicyclopentadiene

purum, ≥95.0% (GC)

Synonym(s):

4,7-Methano-3a,4,7,7a-tetrahydroindene, Cyclopentadiene dimer, DCPD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12
CAS Number:
Molecular Weight:
132.20
Beilstein:
1904092
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥95.0% (GC)

contains

~0.05% 2,6-di-tert.-butyl-4-methylphenol as stabilizer

refractive index

n20/D 1.511

bp

170 °C (lit.)

mp

33 °C (lit.)

density

0.986 g/mL at 25 °C (lit.)

SMILES string

C1C=CC2C3CC(C=C3)C12

InChI

1S/C10H12/c1-2-9-7-4-5-8(6-7)10(9)3-1/h1-2,4-5,7-10H,3,6H2/t7-,8+,9?,10?/m0/s1

InChI key

HECLRDQVFMWTQS-AFWXGSBKSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product has been replaced by 35170-ALDRICH | 2,4-Dichloroaniline ≥97.0% (GC)
Preparation of cyclopentadiene

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R.B. Moffett
Organic Syntheses, IV, 238-238 (1963)
L G Stehmeier et al.
Canadian journal of microbiology, 42(10), 1051-1060 (1996-10-01)
Degradation of dicyclopentadiene (DCPD) to carbon dioxide and oxygenated intermediates was established in the laboratory. Screening of many inocula using BIOLOGMT plates showed that no single colony isolate readily mineralized DCPD. Mixed cultures from a variety of environmental sources produced
Jason M Kamphaus et al.
Journal of the Royal Society, Interface, 5(18), 95-103 (2007-06-21)
Using self-healing materials in commercial applications requires healing chemistry that is cost-effective, widely available and tolerant of moderate temperature excursions. We investigate the use of tungsten (VI) chloride as a catalyst precursor for the ring-opening metathesis polymerization of exo-dicyclopentadiene (exo-DCPD)
E N Brown et al.
Journal of microencapsulation, 20(6), 719-730 (2003-11-05)
Microencapsulated healing agents that possess adequate strength, long shelf-life and excellent bonding to the host material are required for self-healing materials. Urea-formaldehyde microcapsules containing dicyclopentadiene were prepared by in situ polymerization in an oil-in-water emulsion that meet these requirements for
Allergic contact dermatitis from unsaturated polyester resin consisting of maleic anhydride, phthalic anhydride, ethylene glycol and dicyclopentadiene.
Keiko Minamoto et al.
Contact dermatitis, 46(1), 62-63 (2002-03-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service